Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems XL BCR/ABL1/ASS DNA Probe
SDP

Catalog No. 22200345
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-345 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-345 Supplier MetaSystems Supplier No. D5082100TC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

Three probes: Orange, green and blue (aqua)

DNA probe specific for the t(9;22)

  • Orange labeled probe hybridizes to extended region at ABL1 locus on 9q34
  • Green labeled probe hybridizes specifically to extended regions at BCR gene on 22q11
  • Blue (aqua) labeled probe specific for ASS region is added to analyse deletions at 9q34 which can be associated with t(9;22)

Specifications

Concentration 100 μL
Content And Storage Three labeled probes: Orange, green, blue (aqua)
Quantity 10 Tests
For Use With (Application) Genotyping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.