Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems XT EGFR amp DNA Probe
SDP

Catalog No. 22200365
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-365 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-365 Supplier MetaSystems Supplier No. D6005100OG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

Two labeled probes: orange , green

Detects amplifications in short arm of chromosome 7

  • Orange labeled probe hybridizes to EGFR locus at 7p11
  • Green labeled probe hybridizes to 7cen region

Specifications

Concentration 100 μg
Content And Storage Two labeled probes: orange and green
Quantity 10 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.