Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems XT ERG-TMPRSS2 DNA Probe
SDP

Catalog No. 22200347
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-347 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-347 Supplier MetaSystems Supplier No. D6006100TC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

Three labeled probes: Orange, green and blue (aqua)

Detects deletions and translocations involving ERG locus in 21q22

  • Green labeled probe hybridizes to region proximal to ERG locus
  • Orange labeled probe hybridizes specifically to distal region adjacent to ERG gene
  • Blue (aqua) labeled probe detects region distal to the TMPRSS2 region which can be fused to ERG by deletion of sequences between ERG and TMPRSS2
  • For Tissue-FISH applications

Specifications

Concentration 100 μL
Content And Storage Three labeled probes: orange, green, blue
For Use With (Equipment) MetaSystems' pretreatment kit
Quantity 10 Tests
For Use With (Application) Genotyping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.