Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems XT PTEN/GRID1 Locus-Specific Probe
SDP

Catalog No. 22200351
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-351 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-351 Supplier MetaSystems Supplier No. D6007100TC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Three labeled probes: orange, green and blue

Detects deletions in 10q23.

  • Orange probe hybridizes to distal PTEN region
  • Green labeled probe hybridizes to GRID1
  • Blue labeled probe hybridizes to centromere of chromosome 10
  • Used for Tissue-FISH applications

Specifications

Concentration 100 μL
Content And Storage Three labeled probes: orange, green and blue
For Use With (Equipment) MetaSystems' pretreatment kit.
Quantity 10 Tests
For Use With (Application) Genotyping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.