Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meteorin-like/METRNL Antibody (829535), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB66791G
Description
Meteorin-like/METRNL Monoclonal antibody specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Meteorin-like/METRNL | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
284207 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
829535 | |
Flow Cytometry, CyTOF-ready | |
meteorin, glial cell differentiation regulator-like, meteorin-like protein, MGC99788 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse METRNL, Gln46-Glu311, Accession # Q8VE43 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction