Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meteorin-like/METRNL Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Supplier: R&D Systems AF7867SP
This item is not returnable.
View return policy
Description
Meteorin-like/METRNL Polyclonal antibody AF7867SP specifically detects Meteorin-like/METRNL in Human samples. It is validated for Western BlottingSpecifications
Meteorin-like/METRNL | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788 | |
Sheep | |
Affinity Purified | |
RUO | |
284207 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
Q641Q3 | |
METRNL | |
Chinese hamster ovary cell line CHO-derived recombinant human Meteorin-like/METRNL, Gln46-Asp311, Accession # Q641Q3 | |
25 μg | |
Primary | |
Detects human Meteorin-like/METRNL in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Meteorin-like/METRNL is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction