Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Meteorin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562335
Description
Immunogen sequence: VTHDVELQES VITVVAARVL RQTPPLFQAG RSGDQGLTSI RTPLRCGVHP GPGTFLFMGW SRFG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
Meteorin is a gene encoding a protein that is part of the cytokine and growth factor family, primarily involved in neurogenesis and gliogenesis. It is located on chromosome 17p13.1 and is highly expressed in the nervous system, where it plays a role in promoting the differentiation and maturation of astrocytes and neural precursor cells. Meteorin functions by enhancing neurite outgrowth and synaptic development, crucial for proper neural network formation. Additionally, it has been implicated in angiogenesis, contributing to the development and maintenance of blood vessels in neural tissue. Research suggests that Meteorin has neuroprotective properties and may help in recovery from neural injury, making it a molecule of interest for therapeutic strategies in neurodegenerative diseases and brain injuries. Its expression and activity are also being studied in relation to inflammatory responses and potential roles in other tissues, offering insights into its diverse biological functions beyond the central nervous system.
Specifications
Meteorin | |
Polyclonal | |
Unconjugated | |
METRN | |
1810034B16Rik; C16orf23; c380A1.2; hypoxia/reoxygenation regulatory factor; hypoxia/reoxygenation regulatory factor in astrocytes; Hyrac; Meteorin; meteorin, glial cell differentiation regulator; METRN; RGD1306222; RJD6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79006 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJH8 | |
METRN | |
Recombinant protein corresponding to Human Meteorin. Recombinant protein control fragment (Product #RP-101168). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction