Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meteorin Polyclonal Antibody, Abbomax

Catalog No. 502203894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3894 Supplier Abbomax Inc Supplier No. 600430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Brain tissue Cellular location: Extracellular or cytoplasmic.

Meteorin Polyclonal antibody specifically detects Meteorin in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot

Specifications

Antigen Meteorin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene METRN
Gene Accession No. Q5Q0T9, Q8C1Q4, Q9UJH8
Gene Alias 1810034B16Rik; C16orf23; c380A1.2; hypoxia/reoxygenation regulatory factor; hypoxia/reoxygenation regulatory factor in astrocytes; Hyrac; Meteorin; meteorin, glial cell differentiation regulator; METRN; RGD1306222; RJD6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of Meteorin protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287151, 70083, 79006
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.