Learn More
Meteorin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA578574
Description
Positive Control: human heart Predicted Reactivity: Mouse (100%), Rat (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Meteorin regulates glial cell differentiation and promotes the formation of axonal networks during neurogenesis.
Specifications
Meteorin | |
Polyclonal | |
Unconjugated | |
METRN | |
meteorin, glial cell differentiation regulator, C16orf23, c380A1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79006 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.31 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Q9UJH8 | |
METRN | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human METRN. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.