Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Metformin hydrochloride
GREENER_CHOICESDP

Catalog No. 2864100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-641-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-641-00 Supplier R&D Systems™ Supplier No. 2864/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Activator of LKB1/AMPK; antidiabetic agent

Metformin hydrochloride is an antidiabetic agent; lowers plasma glucose levels and improves insulin sensitivity. Inhibits hepatic gluconeogenesis via activation of the LKB1/AMPK pathway. Displays antiproliferative effects in cancer cell lines via inhibition of mitochondrial complex I (MRC-1). Activates the aPKC-CBP pathway in neural precursors to promote neurogenesis. Activates autophagy. Identified by chemoinformatics as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 165.62
Purity >98%
Molecular Weight (g/mol) 165.63
Chemical Name or Material Metformin hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.