Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metformin Hydrochloride, BP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18604343 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

M1566, 1115-70-4, C4H11N5·HCl

Spectrum™ Chemical Metformin Hydrochloride, BP is an antidiabetic used in the treatment of type 2 diabetes. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.5 to 101%
Grade BP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.