Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrolein, 96%, stab. with hydroquinone

Catalog No. p-7058726
Click to view available options
Quantity:
5 mL
25 mL

CAS: 78-85-3 | C4H6O | 70.09 g/mol

Methacrolein is used in the manufacturing of polymers, synthetic resins and plastics. It acts as a raw material of methylmalonic acid, thermoplastic fuel and in the preparation of 2,3-dibromo-2-methyl-propionaldehyde. It is also used to study the effect limonene oxidation products and the terpene oxidation product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-85-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00006974
InChI Key STNJBCKSHOAVAJ-UHFFFAOYSA-N
Synonym methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
PubChem CID 6562
IUPAC Name 2-methylprop-2-enal
SMILES CC(=C)C=O

Specifications

Melting Point -81°C
Color Yellow
Density 0.847 g/mL
Boiling Point 69°C to 70°C
Flash Point −15°C (5°F)
Odor Pungent
Refractive Index 1.415
Linear Formula H2C=C(CH3)CHO
Quantity 5 mL
UN Number UN2396
Sensitivity Light sensitive
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 70.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methacrolein, Stabilized with hydroquinone
Show More Show Less
Hazard Category H225-H301+H311-H314-H330
Hazard Statement GHS H Statement
H225-H300-H310-H330-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHACRYLALDEHYDE, STABILIZED
EINECSNumber 201-150-1
RTECSNumber OZ2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.