Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylamide, 98%

Catalog No. aal1501322
Click to view available options
Quantity:
100 g
500 g

CAS: 79-39-0 | C4H7NO | 85.106 g/mol

Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

Solubility
Soluble in water (202g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Specifications

Melting Point 107°C to 112°C
Density 1.115
Boiling Point 215°C
Quantity 100 g
Beilstein 605397
Solubility Information Soluble in water (202g/L at 20°C).
Formula Weight 85.11
Percent Purity 98%
Chemical Name or Material Methacrylamide
Hazard Category H302-H319-H335-H371-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P309+P311-P312-P330-P501c
EINECSNumber 201-202-3
RTECSNumber UC6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.