Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacryloyl chloride, 95%, contains 200 ppm MEHQ as stabilizer, AcroSeal™, Thermo Scientific™

Catalog No. AC168661000
Click to view available options
Quantity:
25 g
100 mL
1 L
Packaging:
AcroSeal™ Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-46-7
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl

Specifications

Melting Point -60.0°C
Density 1.0760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H5ClO
Refractive Index 1.4447
Linear Formula H2C=C(CH3)COCl
Quantity 100 mL
Beilstein 02,II,394
Specific Gravity 1.076
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common inert organic solvents,like ether,acetone and chloroform
Molecular Weight (g/mol) 104.54
Stabilizer 200ppm MEHQ max.
Formula Weight 104.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methacryloyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
EINECSNumber 213-058-9
RTECSNumber OZ5791000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.