Learn More
Methacryloyl chloride, may contain up to ca. 15% cyclic dimer, 97%, stab., Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methacryloyl chloride is used in the preparation of polymeric materials. It reacts with L-histidine and forms functional monomer. It is also used to prepare monomer 2-methacrylamido-caprolactam, amide monomers by amidation.
Solubility
Miscible with alcohols, ethers and organic solvents. Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -60°C |
| Density | 1.07 |
| Boiling Point | 95°C to 96°C |
| Flash Point | 2°C (35°F) |
| Odor | Acrid |
| Refractive Index | 1.444 |
| Quantity | 100 g |
| UN Number | UN3488 |
| Beilstein | 878175 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H314-H335 |
| Hazard Statement | GHS H Statement H225-H330-H314-H318-H302 Highly flammable liquid and vapor. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, CORROSIVE, N.O.S. |
| EINECSNumber | 213-058-9 |
| RTECSNumber | OZ5791000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.