Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Methamphetamine-D9 solution, 1 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant

Catalog No. M0911ML Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mL

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes.

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes. This internal standard is suitable for quantitation of methamphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, sports testing, forensic analysis, or isotope dilution methods. The dextrorotatory isomer of methamphetamine is a pharmaceutical drug sold under the trade name Desoxyn™ for treatment of ADHD and obesity.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Methamphetamine-D9
Packaging Ampule of 1 mL
CAS 169565-19-9
Molecular Formula C10D9H6N
Concentration 1 mg/mL in Methanol (as free base)
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 158.29
Quantity 1 mL
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.