Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+)-Methamphetamine hydrochloride Solution, 1.0 mg/mL in methanol, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 mL
Description
Featured Industries: Pharmaceutical (small molecule)

Specifications
Specifications
CAS | 51-57-0 |
Flash Point | 9°C |
Concentration | 1.0 mg/mL in methanol |
Linear Formula | C10H15 N · HCl |
MDL Number | MFCD00056130 |
Type | Analytical Standard |
Industry Type | Drug Analysis |
Quantity | 1 mL |
Formula Weight | 185.69 |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction