Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Methandriol Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175275
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175275 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175275 Supplier Invitrogen™ Supplier No. PA175275
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 10 μg/mL. Sensitivity: 10 ng/mL Methandriol produces 31% inhibition in a competitive ELISA employing Methandriol polyclonal antibody.

Methandriol is a synthetic steroid with anabolic and androgenic properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methandriol
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.8
Gene Alias C20H32O2; Diolandrone; Mestenediol; Metandriol; Methandrolan; Methylandrostenediol
Host Species Sheep
Immunogen Methandriol(3b)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.