Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanephosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC354330250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC354330250 25 g
AC354330050 5 g
AC354331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC354330250 Supplier Thermo Scientific Chemicals Supplier No. 354330250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-13-5
Molecular Formula CH5O3P
Molecular Weight (g/mol) 96.02
MDL Number MFCD00002137
InChI Key YACKEPLHDIMKIO-UHFFFAOYSA-N
Synonym methanephosphonic acid, phosphonic acid, methyl, methyl phosphonic acid, unii-329w4ym10z, phosphonomethyl group, ester group, phosphonomethyl, acmc-20ake9, d02fxw, methylphosphonic acid
PubChem CID 13818
ChEBI CHEBI:45129
IUPAC Name methylphosphonic acid
SMILES CP(O)(O)=O

Specifications

Melting Point 103.0°C to 107.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 96.02
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methanephosphonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 213-607-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.