Learn More
Methanesulfonic acid, 70% aq. soln.
Description
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.
Solubility
Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.
Notes
Light and heat sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.
Specifications
Specifications
| Density | 1.32 |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.415 |
| Quantity | 2500 mL |
| UN Number | UN2586 |
| Merck Index | 14,5954 |
| Solubility Information | Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins. |
| Formula Weight | 96.1 |
| Concentration or Composition (by Analyte or Components) | 70% aq. soln. |
| Chemical Name or Material | Methanesulfonic acid |
Safety and Handling
| Hazard Category | H290-H302+H312-H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 200-898-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.