Learn More
R&D Systems™ Methanesulfonic Acid Stop Solution, 16 x 12 mL vials
Shop All Novus Biologicals Products

Specifications
Specifications
| Quantity | 1 Pack |
| Binding Target | Buffers, Chelators and Reagents |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.