Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Methanesulfonic Acid Stop Solution, 250 mL
SDP

Catalog No. DY994B250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
250 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY994B250 250 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DY994B250 Supplier R&D Systems™ Supplier No. DY994B250

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Methanesulfonic Acid Stop Solution, 250 mL

Specifications

Quantity 250 Pack
Binding Target Buffers, Chelators and Reagents
Product Line R&D Systems
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.