Learn More
Methanesulfonyl chloride, 98%
Description
Methanesulfonyl chloride is used as a reagent for conversion of alcohols to mesylate esters such as methanesulfonate, which is an intermediate in substitution reactions, elimination reactions, reductions, and rearrangement reactions viz. Beckmann rearrangement. It is an electrophile and acts as a source of CH3SO2+ group. It is also used to prepare beta-chloro sulfones, methanesulfonamide and heterocyclic compounds containing five membered sultones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methanesulfonyl chloride is used as a reagent for conversion of alcohols to mesylate esters such as methanesulfonate, which is an intermediate in substitution reactions, elimination reactions, reductions, and rearrangement reactions viz. Beckmann rearrangement. It is an electrophile and acts as a source of CH3SO2+ group. It is also used to prepare beta-chloro sulfones, methanesulfonamide and heterocyclic compounds containing five membered sultones.
Solubility
Miscible with alcohol, ether and organic solvents. Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong bases, oxidizing agents and alcohols. It is a lachrymator and reacts with water.
Specifications
Specifications
| Melting Point | °C to 33°C |
| Density | 1.477 |
| Boiling Point | 161°C |
| Flash Point | 97°C (206°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.452 |
| Quantity | 100 g |
| UN Number | UN3246 |
| Beilstein | 506297 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H290-H301+H311-H314-H317-H330-H335 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318 |
| Precautionary Statement | P234-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHANESULFONYL CHLORIDE |
| EINECSNumber | 204-706-1 |
| RTECSNumber | PB2790000 |
| TSCA | Yes |
| storageNote1 | Moisture Sensitive |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.