Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol with 0.1% acetic acid, Solstice

Catalog No. 60090271 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-271 4 x 2.5 L
60-090-292 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-271 Supplier Solstice Supplier No. 346724X2.5L
Add to Cart
Add to Cart

CHROMASOLV™ LC-MS, LabReady™ Solvent Blend

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1 , 64-19-7
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 99.90%, 0.10%
Quantity 4 x 2.5 L
Flash Point 20°C
Packaging Glass Bottle
Physical Form Liquid, clear
Boiling Point 68°C
Grade LC-MS solvent blends
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.