Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol with 0.1% Formic Acid, Solstice

Catalog No. 60044847 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
60-044-847 Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-847 Supplier Solstice Supplier No. 142762L

missing translation for 'validMainframeMsg'

CHROMASOLV™ LC-MS Ultra, LabReady™ Solvent Blend, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6 , 67-56-1
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
MDL Number MFCD00004595
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
SMILES OC=O

Specifications

Packaging Glass Bottle
UN Number UN1230
Formula Weight 32.04g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Methanol With 0.1% Formic Acid
Hazard Statement P210-P260-P280-P301 + P310 + P330-P308 + P311-P403 + P233
Precautionary Statement H225-H301 + H311 + H331-H370
DOTInformation class 3 - PG 2 - Methanol, solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.