Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, 77% v/v, Reagents
Click to view available options
:
Quantity: 4 L
Specifications
Specifications
Color | Colorless |
Boiling Point | 68°C |
Quantity | 4 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Specific Gravity | 0.8657 |
Percent Purity | 77% |
Grade | Reagent |
Chemical Name or Material | Methanol, 77% v/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction