Learn More
Methanol, Absolute, Reagent, ACS, 99.8%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Methanol, Absolute, Reagent, ACS , is also called methyl alcohol or 'wood alcohol' it was one of the by products of distillation of wood. As the simplest alcohol around, it is light, flammable, colorless liquid with a strong and distinctive odor. It has many uses including acting as an antifreeze, (due to it's polar liquid properties), solvent, fuel as well as a denaturant for ethanol. As an ACS grade Reagent, Spectrum Chemical manufactured methanol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 6 x 1 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.001% |
| UN Number | UN1230 |
| Boiling Point | 64°C to 65°C |
| Percent Purity | 99.8% |
| Grade | Reagent |
| Chemical Name or Material | Methanol |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.