Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents

Catalog No. 01338140
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
Poly Bottle
Steel Drum
Steel Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-140 1 L Glass Bottle
01-338-141 10 L Steel Pail
01-338-139 2.5 L Glass Bottle
01-338-138 200 L Steel Drum
01-338-137 20 L Steel Pail
01-338-136 4 L Poly Bottle
01-338-135 4 L Glass Bottle
01-338-134 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-338-140 Supplier Reagents Holdings Llc Supplier No. C11210001C
Add to Cart
Add to Cart

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7866
Boiling Point 64.5°C
Grade ACS
Chemical Name or Material Methanol, ACS Reagent, Absolute (Methyl Alcohol, Anhydrous)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.