Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
SDP

Catalog No. R482900020E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Hard Pack
Metal Can
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R482900020E 20 L Metal Can
R482900025C 25 mL Amber Glass Bottle
48-291-6 500 mL Natural Poly Bottle
482932 1 L Natural Poly Bottle
48291 4 L Natural Poly Bottle
48295HP 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R482900020E Supplier Ricca Chemical Company Supplier No. R482900020E4
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Quantity 20 L
Packaging Metal Can
Physical Form Liquid
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0003 meq/g
Water max. 0.1%
Synonym methyl alcohol
Boiling Point 64.5°C
Percent Purity min. 99.8%
Grade ACS
Chemical Name or Material Methanol, Absolute
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.