Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Solstice

Catalog No. 6000978 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-009-78 19 L Metal Can
60-009-80 4 x 4 L Glass Bottle
60-009-81 4 x 4 L Poly Bottle
60-009-79 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-009-78 Supplier Solstice Supplier No. LP23019
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 19 L
Flash Point 11°C
Packaging Metal Can
Water 0.1% max.
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.8%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Methanol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.