Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Purge and Trap grade, for VOC Analysis using Purge and Trap methods, >99.9%, Solstice

Catalog No. 6000984 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
12 x 235 mL
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-009-84 12 x 235 mL Glass Bottle
60-009-85 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-84 Supplier Solstice Supplier No. 232235
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 12 x 235 mL
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity >99.9%
Grade Purge and Trap
Chemical Name or Material Methanol, B&J Brand™, Purge & Trap grade, for VOC Analysis using Purge & Trap methods, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.