Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, B&J Brand™, Purge & Trap grade, for VOC Analysis using Purge & Trap methods, >99.9%, Honeywell Burdick & Jackson

Catalog No. p-7111767
Click to view available options
Quantity:
12 x 235 mL
6 x 1 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Boiling Point 64.7°C
Quantity 12 x 235 mL
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04
Percent Purity >99.9%
Grade Purge and Trap
Chemical Name or Material Methanol, B&J Brand™, Purge & Trap grade, for VOC Analysis using Purge & Trap methods, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.