Learn More
Methanol, BioReagent, ≥99.93%, For Protein Sequencing,Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Solstice developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications
Specifications
| Melting Point | -98°C |
| Density | 0.791g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.36 |
| Flash Point | 9°C (48.2°F) |
| Ignition Point | 725°F |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 410mmHg (50°C), 97.68mmHg (20°C) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235 |
| DOTInformation | class 3 - PG 2 - Methanol |
| RTECSNumber | 200-659-6 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.