Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Capillary GC Grade; ≥99.9%, Solstice

Catalog No. 60046834 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-834 4 L Glass Bottle
60-046-835 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-834 Supplier Solstice Supplier No. 4147194L
Add to Cart
Add to Cart

Capillary GC grade,≥99.9%

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.791g/mL at 25°C
Quantity 4 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.329
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Capillary GC
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
DOTInformation class 3 - PG 2 - Methanol
RTECSNumber 200-659-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.