Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d4,

Catalog No. AC464040500
Click to view available options
Quantity:
1 mL
10 mL
25 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CD4O | 36.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CD4O
Molecular Weight (g/mol) 36.06

Specifications

Density 0.8880 g/mL
Quantity 50 mL
Refractive Index 1.3262
Flash Point 12°C
Specific Gravity 0.8880
Melting Point -99.0°C
Boiling Point 65.0°C
CAS 811-98-3
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Grade NMR
Chemical Name or Material Methanol-d4
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes damage to organs.

Toxic if swallowed.

Toxic in contact with skin.

Toxic if inhaled.

Highly flammable liquid and vapor.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower Wash face, hands and any exposed skin thoroughly after handling.

Wear protective gloves/protective clothing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician.

EINECSNumber 212-378-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.