Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Solstice

Catalog No. 60090370 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-370 200 L
60-090-378 56 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-370 Supplier Solstice Supplier No. BC233200
Add to Cart
Add to Cart

BioSyn™, for biosynthesis and applications requiring low water content, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
PubChem CID 887
ChEBI CHEBI:17790

Specifications

Quantity 200 L
Flash Point 11 °C
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.