Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Lab Grade), Fisher Science Education™ PROMO

Catalog No. S25426AS24 Shop All Fisherbrand Products
Change view
Click to view available options
:
Methanol; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S25426AS24 Methanol; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25426AS24

Synonym: Methyl Alcohol, Wood Alcohol, Carbinol

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Density 0.791 g/mL
Quantity 4 L
Linear Formula CH3OH
Formula Weight 32.04
Chemical Name or Material Methanol
ShelfLife 2 Years
DOTInformation DOT Class 3, II: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.