Alfa Aesar™ Methanol, LC-MS Grade, ≥99.8%

Solvent for LC-MS analysis

Manufacturer: Alfa Aesar™ 47192K2

Quantity 1L

 View more versions of this product

Catalog No. AA47192K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-56-1
    Linear Formula CH3OH
    Formula Weight 32.04
    MDL Number MFCD00004595
    UN Number UN1230
    Beilstein 1098229
    Merck Index 14,5957
    Chemical Name or Material Methanol
    Grade LC/MS Grade

    Specifications

    Formula Weight 32.04
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1230
    Health Hazard 2 GHS H Statement
    H225-H301-H311-H331-H370
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
    Odor Mild
    Refractive Index 1.3290
    Boiling Point 64.7°C
    Color Colorless
    Quantity 1L
    Linear Formula CH3OH
    CAS 67-56-1
    Beilstein 1098229
    Merck Index 14,5957
    Health Hazard 1 Danger
    Density 0.791g/mL
    RTECS Number PC1400000
    TSCA TSCA
    Melting Point −98°C
    MDL Number MFCD00004595
    Physical Form Liquid
    Chemical Name or Material Methanol
    Assay ≥99.8%
    Grade LC/MS Grade

    GHS H Statement
    H225-H301-H311-H331-H370
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit