Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Solstice

Catalog No. 60046086 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
45 kg
4 x 5 L
6 x 1 L
150 kg
Packaging:
Metal Drum
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-086 45 kg Metal Drum
60-046-089 5 L Poly Bottle
60-046-090 6 x 1 L Poly Bottle
60-046-087 4 x 2.5 L Poly Bottle
60-046-088 4 x 5 L Poly Bottle
60-046-082 150 kg Metal Drum
60-046-083 1 L Poly Bottle
60-046-084 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60-046-086 Supplier Solstice Supplier No. 3221345KG
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Appearance Liquid
Density 0.791g/mL at 25°C
Quantity 45 kg
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Metal Drum
Refractive Index 1.328 to 1.33
Residue after Evaporation 0 to 0.0005 %
Titratable Acid Free acid (as HCOOH): ≤0.002%
Titratable Base Free Alkali (as NH3): ≤0.00005%
Linear Formula CH3OH
Vapor Pressure 410mmHg (50°C), 97.68mmHg (20°C)
UN Number UN1230
Beilstein 1098229
Boiling Point 64°C to 65°C
Formula Weight 32.04g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Methanol, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Statement P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
Precautionary Statement H225-H301 + H311 + H331-H370
DOTInformation Class 3 - PG 2 - Methanol
EINECSNumber 200-659-6
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.