Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Methaqualone Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175279
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175279 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175279 Supplier Invitrogen™ Supplier No. PA175279
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 1/4K. Sensitivity: 10 ng/mL Methaqualone Sigma M3657 produces 41% inhibition in a competitive ELISA employing Methaqualone polyclonal antibody.

Methaqualone is a quinazoline derivative that acts as a sedative and hypnotic agent. It was been previously used for insomnia but withdrawn from the market in many countries because of problems with abuse. [National Center for Biotechnology Information. PubChem Compound Database; CID=6292].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methaqualone
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.9
Gene Alias C16H14N2O; Cateudyl; Metaqualon; Metolquizolone; Ortonal; Quaalude
Host Species Sheep
Immunogen Methaqualone(6')-BSA.
Purification Method purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.