Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methicilin-resistant Staph aureus (MRSA) Monoclonal Antibody (ANT-198), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 15715500UG

This item is not returnable.
View return policy
Description
Methicilin-resistant Staph aureus (MRSA) Monoclonal antibody specifically detects Methicilin-resistant Staph aureus (MRSA) in Bacteria samples. It is validated for ELISA, Western BlotSpecifications
Methicilin-resistant Staph aureus (MRSA) | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
ANT-198 | |
Unconjugated | |
MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant | |
Gel-purified MRSA protein from strain EMRSA 16 | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction