Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methicillin-resistant Staphylococcus aureus Mouse anti-Bacteria, Unconjugated, Clone: HM504, Abnova™

Catalog No. 89339320 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-320 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-320 Supplier Abnova Corporation Supplier No. MAB21934

Mouse Monoclonal Antibody.

Methicillin-resistant Staphylococcus aureus Monoclonal antibody specifically detects Methicillin-resistant Staphylococcus aureus in Bacteria samples. It is validated for WB, IP, ELISA, IHC.

Specifications

Antigen Methicillin-resistant Staphylococcus aureus
Applications Western Blot, Immunoprecipitation, ELISA, Immunohistochemistry
Classification Monoclonal
Clone HM504
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified MRSA protein (mecA)
Quantity 1 mg
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 1 mL deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.