Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methicillin Sodium Salt, 90+%, PROMO, Thermo Scientific Chemicals

Catalog No. J6641403PM
Click to view available options
Quantity:
1 g

Applications

Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP's are enzymes which catalyze a pentagl

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-92-3
Molecular Formula C17H19N2NaO6S
Molecular Weight (g/mol) 402.4 g/mol
MDL Number MFCD07787409
InChI Key MGFZNWDWOKASQZ-UMLIZJHQSA-M
Synonym Sodium (2,6-dimethoxyphenyl)penicillin; Sodium methicillin
PubChem CID 23667627
ChEBI CHEBI:52065
IUPAC Name sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O

Specifications

Type Methicillin Sodium Salt
Physical Form Powder
Melting Point 196°C to 197°C (decomposition)
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Sensitivity Light Sensitive
Boiling Point 640°C
Merck Index 14,5969
Solubility Information Slightly soluble in acetone; Soluble in ethanol or water
Percent Purity 98%
Chemical Name or Material Methicillin sodium salt
Show More Show Less
EINECSNumber 205-083-9
RTECSNumber XH8950000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.