Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299658AF405
Description
Methionine Aminopeptidase 1D/MAP1D Polyclonal specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISA.Specifications
Methionine Aminopeptidase 1D/MAP1D | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 405 | |
EC 3.4.11.18, MAP1DCDS of metAP-3 within PCR fragment, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial | |
Produced in rabbits immunized with purified, recombinant Methionine Aminopeptidase 1D/MAP1D (NP_954697.1; Arg44-Ala335). | |
0.1 mL | |
Immunology | |
254042 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction