Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Rabbit anti-Human, Clone: 8, Novus Biologicals™

Rabbit Monoclonal Antibody
$345.19 - $554.59
Specifications
| Antigen | Methionine Aminopeptidase 1D/MAP1D |
|---|---|
| Clone | 8 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Methionine Aminopeptidase 1D/MAP1D Monoclonal specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISA.Specifications
| Methionine Aminopeptidase 1D/MAP1D | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 254042 | |
| IgG | |
| Protein A purified |
| 8 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 3.4.11.18, MAP1DCDS of metAP-3 within PCR fragment, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial | |
| Recombinant Human Methionine Aminopeptidase 1D/MAP1D protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title