Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methionine Aminopeptidase 2/METAP2 Antibody (001), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290622MFV500
Description
Methionine Aminopeptidase 2/METAP2 Monoclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Methionine Aminopeptidase 2/METAP2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
001 | |
Immunohistochemistry-Paraffin | |
eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2 | |
Rabbit immunized with purified, recombinant Mouse Methionine Aminopeptidase 2/METAP2 (Uniprot#: O08663, Ala2-Tyr478) | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
10988 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction