Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methionine Aminopeptidase 2/METAP2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299115AF700
Description
Methionine Aminopeptidase 2/METAP2 Polyclonal specifically detects Methionine Aminopeptidase 2/METAP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Methionine Aminopeptidase 2/METAP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
10988 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2 | |
Produced in rabbits immunized with purified, recombinant Methionine Aminopeptidase 2/METAP2 (NP_006829.1; Met1-Tyr478). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction