Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methionine Aminopeptidase 2/METAP2 Rabbit anti-Human, Mouse, Rat, Clone: 5U2F4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Methionine Aminopeptidase 2/METAP2 |
---|---|
Clone | 5U2F4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Methionine Aminopeptidase 2/METAP2 Monoclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Methionine Aminopeptidase 2/METAP2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10988 | |
IgG | |
Affinity purified |
5U2F4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 380-478 of human Methionine Aminopeptidase 2/METAP2 (P50579). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title