Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methocarbamol, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604392 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

M1088, 532-03-6, C11H15NO5

Spectrum™ Chemical Methocarbamol, USP is used to treat muscle spasms and pain. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-03-6
Molecular Formula C11H15NO5
Molecular Weight (g/mol) 241.24
InChI Key GNXFOGHNGIVQEH-UHFFFAOYNA-N
IUPAC Name 2-hydroxy-3-(2-methoxyphenoxy)propyl carbamate
SMILES COC1=CC=CC=C1OCC(O)COC(N)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98.5 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.