Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothall Standard from Endothall Monohydrate, SPEX CertiPrep™
SDP

Catalog No. 5481A
Click to view available options
Quantity:
1 mL

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Endothal Monohydrate62059-43-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 62059-43-2, 67-56-1
Molecular Formula C8H12O6
Molecular Weight (g/mol) 204.178
InChI Key RHLALKDQINFLPM-UHFFFAOYSA-N
PubChem CID 23615703
IUPAC Name 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid;hydrate
SMILES C1CC2C(C(C1O2)C(=O)O)C(=O)O.O

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.2, 99.8
CAS Max % 0.2, 99.8
Methods For US EPA Method 548.1
Analyte or Component Names Endothal
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element Endothal
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material Endothall (from Endothall Monohydrate)
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.